Labels, hazard warning
Hazard labels: danger, auto, —, Polyester, zwart op geel, 50×43 mm, 7 labels/card
Catalogus # 116-0755
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Specificaties
- Kleur:zwart op geel
- Omschrijving:Hazard labels: danger, auto
- Afmetingen:50×43 mm
- Materiaal:Polyester
- Seq:13
- Model:—
- Verpakt:7 labels/card