[EN]CAPILLARIES ON BOTH SIDE OPEN 1 * 100 ST
BUCH040807
: BUCHI
Frequently Bought Together
Bekerglazen, laag model, DURAN®
: DWK Life Sciences
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Patroon 220G voor patroonbrander
: USBECK Laborgeräte
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Accessoires voor mechanische pipetten
: VWR Collection
Precisiethermometers
: Amarell