Funnels, powder
50 mm
Catalogus # WITG5.790.001
Leverancier: witeg Labortechnik
Ga naar:
Product Details & Documents
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specificaties
- Trechter uitw. Ø:50 mm