Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Funnels, powder
50 mm
Catalogus # WITG5.790.001
Leverancier:   witeg Labortechnik
Funnels, powder
Catalogus # WITG5.790.001
Leverancier:  witeg Labortechnik
Funnels, powder
Funnels, powder
50 mm
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Powder funnels, made of DURAN® borosilicate glass 3.3, with short wide stem.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specificaties
  • Trechter uitw. Ø:
    50 mm