Lempert rongeurs
Lempert rongeurs, 2,5 mm cup diameter, Curved, 160 mm
FSCI16005-16
Marketsource
These rongeurs are made of stainless steel.
- Single joint action
- Type:Curved
- Omschrijving:Lempert rongeurs, 2,5 mm cup diameter
- Lengte:160 mm
- Seq:02