Potts-Smith forceps
Roestvrij staal, 250 mm, Serrated, Straight, 2×1,6 mm
Catalog # FSCI11012-25
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
Specifications
- Lengte:250 mm
- Materiaal:Roestvrij staal
- Seq:07
- Tip stijl:Serrated
- Tipgrootte:2×1,6 mm
- Tip shape:Straight