Stevens Tenotomy scissors
Sharp/sharp, straight tips, 110 mm, 21 mm
Catalogus # FSCI14064-11
Marketsource
Leverancier: FINE SCIENCE TOOLS
Ga naar:
Product Details & Documents
These scissors are made of stainless steel.
Specificaties
- Type:Sharp/sharp, straight tips
- Lengte:110 mm
- Seq:04
- Lengte blad:21 mm