Mayo-Stille scissors
Blunt/blunt, straight tips, 150 mm, 39 mm
Catalog # FSCI14012-15
Marketsource
Supplier: FINE SCIENCE TOOLS
Jump to:
Product Details & Documents
These scissors are made of stainless steel.
Documents
Certificates
Search by Lot NumberSpecifications
- Type:Blunt/blunt, straight tips
- Lengte:150 mm
- Seq:02
- Lengte blad:39 mm