Mayo-Stille scissors
Blunt/blunt, curved tips, 150 mm, 39 mm
Catalogus # FSCI14013-15
Marketsource
Leverancier: FINE SCIENCE TOOLS
Ga naar:
Product Details & Documents
These scissors are made of stainless steel.
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Type:Blunt/blunt, curved tips
- Lengte:150 mm
- Seq:01
- Lengte blad:39 mm