Orders LinkContactUsLinkComponent
Dumont #5 forceps
Dumont #5 forceps
#5, Titaan, 110 mm, Biology, Straight, 0,05×0,02 mm
  FSCI11252-40
Marketsource
Dumont #5 forceps
  FSCI11252-40
Marketsource
Dumont #5 forceps
Dumont #5 forceps
#5, Titaan, 110 mm, Biology, Straight, 0,05×0,02 mm
46,50 €
1 * 1 ST
  • Type:
    #5
  • Lengte:
    110 mm
  • Materiaal:
    Titaan
  • Seq:
    04
  • Tip stijl:
    Biology
  • Tipgrootte:
    0,05×0,02 mm
  • Tip shape:
    Straight