Dumont forceps, micro-blunted tips
#5, Inox, 115 mm, Standard, off-center, Straight, 0,13×0,08 mm
Catalogus # FSCI11253-21
Marketsource
Leverancier: FINE SCIENCE TOOLS
Dumont forceps, micro-blunted tips
Catalogus # FSCI11253-21
Leverancier: FINE SCIENCE TOOLS
Marketsource
Ga naar:
Product Details & Documents
Specificaties
- Type:#5
- Lengte:115 mm
- Materiaal:Inox
- Seq:04
- Tip stijl:Standard, off-center
- Tipgrootte:0,13×0,08 mm
- Tip shape:Straight