Orders LinkContactUsLinkComponent
Dumont forceps, micro-blunted tips
Dumont forceps, micro-blunted tips
#5/45, Inox, 110 mm, Standard, Angled (45°), 0,1×0,06 mm
Catalogus # FSCI11253-25
Marketsource
Leverancier:   FINE SCIENCE TOOLS
Dumont forceps, micro-blunted tips
Catalogus # FSCI11253-25
Leverancier:  FINE SCIENCE TOOLS
Marketsource
Dumont forceps, micro-blunted tips
Dumont forceps, micro-blunted tips
#5/45, Inox, 110 mm, Standard, Angled (45°), 0,1×0,06 mm
Price
48,00 €
Log in voor beschikbaarheid
1 * 1 Pce
Product Details & Documents
Specificaties
  • Type:
    #5/45
  • Lengte:
    110 mm
  • Materiaal:
    Inox
  • Seq:
    02
  • Tip stijl:
    Standard
  • Tipgrootte:
    0,1×0,06 mm
  • Tip shape:
    Angled (45°)