Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Razor blades, Gem®
Razor blades, Gem®
Eenzijdige scheermesjes, Gem® Scientific, 3 facetten, aluminium achterzijde, 100 mesjes per dispenser, Koolstofstaal, 0,22 mm, 38×19 mm
Catalog # PERS62-0325
Supplier:   PERSONNA MEDICAL
Razor blades, Gem®
Catalog # PERS62-0325
Supplier:  PERSONNA MEDICAL
Razor blades, Gem®
Eenzijdige scheermesjes, Gem® Scientific, 3 facetten, aluminium achterzijde, 100 mesjes per dispenser, Koolstofstaal, 0,22 mm, 38×19 mm
Price
1 960,00 €
Log in for availability
1 * 5.000 ST
Product Details & Documents
Razor blades for catheter cutting or cleanroom applications.
  • High precision, sharpness and safety
  • Made from carbon steel or from stainless steel
Carbon steel blades are suitable for ISO Class 7 cleanroom environments.
Stainless steel blades are suitable for ISO Class 5, 6 and 7 cleanroom environments.
Specifications
  • Omschrijving:
    Eenzijdige scheermesjes, Gem® Scientific, 3 facetten, aluminium achterzijde, 100 mesjes per dispenser
  • l×b:
    38×19 mm
  • Materiaal:
    Koolstofstaal
  • Dikte:
    0,22 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.