Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
 

Acetonitril, Prepsolv® for preparative chromatography

Glass bottle, 2,5 L
Catalogus # 1.13358.2509
Ontvlambaar
Irriterend
   Veiligere gezondheid van mens en milieu
Leverancier:   Merck
Acetonitril, Prepsolv® for preparative chromatography
Catalogus # 1.13358.2509
Leverancier:  Merck
   Veiligere gezondheid van mens en milieu
CAS-nummer:  75-05-8
Acetonitril, Prepsolv® for preparative chromatography
Acetonitril, Prepsolv® for preparative chromatography

Glass bottle, 2,5 L

CAS-nummer:  75-05-8
Price
3 420,00 €
Log in voor beschikbaarheid
1 * 2,5 L
Product Details & Documents

Prepsolv® solvents are tailored to the requirements of preparative HPLC to facilitate scale-up from analytical to preparative separations. With their extremely low evaporation residue (< 1 mg/l) and low water content, they ensure optimal protection for columns.

Synoniemen:  Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
  • CAS-nummer:75-05-8
  • Formule:H₃CCN
  • MDL-nummer:MFCD00001878
  • Molecular Weight:41.05 g/mol
  • Merck index:13,00071
  • Kokend Pt:81,6 °C (1013 hPa)
  • Smelten Pt:–45,7 °C
  • Dichtheid:0,782 g/cm³ (20 °C)
  • Flits Pt:2 °C (gesloten beker)
  • UN:1648
  • ADR:3,
  •  :M-Clarity™ quality level = MQ200
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-835-2
  • BEREIKEN:01-2119471307-38

Documents

Certificaten
Zoeken op lotnummer
Specificaties

Specificaties

  • Verpakking:
    Glass bottle
  • VPE:
    2,5 L
  • Cat. No.:
    1.13358.2509
  • Milieu certificering:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Veiligere gezondheid van mens en milieu

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.