Orders LinkContactUsLinkComponent
Washers, BOLA
Washers, BOLA
M 5 thread, 10 mm
Catalogus # BOHLH1126-18
Leverancier:   Bohlender
Washers, BOLA
Catalogus # BOHLH1126-18
Leverancier:  Bohlender
Washers, BOLA
Washers, BOLA
M 5 thread, 10 mm
Price
4,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
PTFE, white.
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • FDA conform
  • Similar to DIN 125-1
Specificaties
  • Ø:
    10 mm
  • Omschrijving:
    M 5 thread