Washers, BOLA
M 8 thread, 16 mm
Catalogus # BOHLH1126-26
Leverancier: Bohlender
Ga naar:
Product Details & Documents
PTFE, white.
- Temperature resistance: from −200 to +250 °C
- Universal chemical resistance
- FDA conform
- Similar to DIN 125-1
Specificaties
- Ø:16 mm
- Omschrijving:M 8 thread