Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Control valves, BOLA
Control valves, BOLA
Control valve with thread M 18×2, for tubing outer Ø 10 mm, PTFE
Catalogus # BOHLE672-56
Leverancier:   Bohlender
Control valves, BOLA
Catalogus # BOHLE672-56
Leverancier:  Bohlender
Control valves, BOLA
Control valves, BOLA
Control valve with thread M 18×2, for tubing outer Ø 10 mm, PTFE
Price
597,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Body made of PTFE with 2 connections and nuts made of glass-fibre reinforced PTFE for connecting tubing or tube.
  • Volume flow can be regulated manually
  • Suitable for panel mounting
  • Easy disassembly for cleaning
  • Universal chemical resistance, the flowing product is only exposed to PTFE
Specificaties
  • Omschrijving:
    Control valve with thread M 18×2, for tubing outer Ø 10 mm, PTFE
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.