Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
1250 ml, 117 mm, 149 mm
Catalogus # SEMA5548
Leverancier:   SEMADENI
Jars with screw cap
Catalogus # SEMA5548
Leverancier:  SEMADENI
Jars with screw cap
Jars with screw cap
1250 ml, 117 mm, 149 mm
Price
558,00 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Jars, made of PP, with tight screw cap.
Suitable for powders, granules, pastes, ointments, waxes, cosmetics and dermatology products.
Specificaties
  • Ø uitw.:
    117 mm
  • Capaciteit:
    1250 ml
  • Hoogte:
    149 mm