Micro Tubes with Thread
Micro tubes without skirted base, neutral cap, 2,0 ml, 1000/case
Catalogus # SARS72.693.465
Marketsource
Leverancier: SARSTEDT
Ga naar:
Product Details & Documents
PP, translucent with PP screw cap.
- With printed label
- Withstand centrifugation up to 20000×g
- Attached, not screwed-on cap
- Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.
Specificaties
- Verpakt:1000/case
- Capaciteit:2,0 ml
- Omschrijving:Micro tubes without skirted base, neutral cap
- Seq:2