Cover slides
24×40 mm, 0,15 mm
Catalog # DELTD102440
Supplier: DELTALAB
Jump to:
Product Details & Documents
Clear and pre-washed glass cover slides dedicated to sample and lens protection.
Delivery information: Supplied in cases of one or two hundred.
Documents
Certificates
Search by Lot NumberSpecifications
- l×b:24×40 mm
- Seq:01
- Dikte:0,15 mm