Dressing forceps, Cushing type
Type Cushing, TC Gold (hard metalen inzetstuk), Recht, stomp, 175 mm
Catalogus # MART12-254-17
Leverancier: KLS Martin Group
Ga naar:
Product Details & Documents
Stainless steel forceps.
- TC Gold: Tungsten Carbide inlays provide an exceptionally long service life of the instruments
Specificaties
- Type:Recht, stomp
- Omschrijving:Type Cushing, TC Gold (hard metalen inzetstuk)
- Lengte:175 mm