Orders LinkContactUsLinkComponent
Accessories for TW alpha plus
Accessories for TW alpha plus
Titration head, TZ 1469, for 24 sample rack TZ 1454, with four openings (2× NS 14,5/2× NS 7,5) and one adapter for micro electrodes (Ø 6 mm)
Catalogus # SCOI285213884
Leverancier:   SI Analytics
Accessories for TW alpha plus
Catalogus # SCOI285213884
Leverancier:  SI Analytics
Accessories for TW alpha plus
Titration head, TZ 1469, for 24 sample rack TZ 1454, with four openings (2× NS 14,5/2× NS 7,5) and one adapter for micro electrodes (Ø 6 mm)
Price
938,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Titration head, TZ 1469, for 24 sample rack TZ 1454, with four openings (2× NS 14,5/2× NS 7,5) and one adapter for micro electrodes (Ø 6 mm)