Dip slides
Total/Coliform + Neutr, 120
Catalogus # LIOF53562
Leverancier: LIOFILCHEM
Ga naar:
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
- ISO 18593
Documents
Certificaten
Zoeken op lotnummerSpecificaties
Specificaties
- VPE:120
- Omschrijving:Total/Coliform + Neutr