Druppelflessen, Lucas
50 ml
Catalog # 215-3583
Supplier: MUELLER KREMPEL
Jump to:
Product Details & Documents
Amber glass, without closure.
- Good chemical and physical properties
- Use in applications that do not involve high thermal stresses
- Please order screw caps and drop dispensers separately
Bruingekleurd ter bescherming van lichtgevoelige materialen.
Specifications
- Capaciteit:50 ml
- Seq:08