Closures for headspace vials
White PTFE coated silicone (natural) cap/septa/spring
Catalogus # PERCN9303992
Leverancier: PerkinElmer
Ga naar:
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specificaties
- Omschrijving:White PTFE coated silicone (natural) cap/septa/spring
- Seq:26