Rectangular spacers for SpectrumGX
Assorted spacers for semi-demountable cell, includes two each of thickness 0,025, 0,05, 0,1, 0,2, 0,5, 1,0 mm spacers, SpectrumGX
Catalogus # PERCL1270421
Leverancier: PerkinElmer
Ga naar:
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specificaties
- Voor:SpectrumGX
- Omschrijving:Assorted spacers for semi-demountable cell, includes two each of thickness 0,025, 0,05, 0,1, 0,2, 0,5, 1,0 mm spacers
- Seq:07