Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 0,5 mm thickness for semi-demountable cell, SpectrumGX
Catalogus # PERCL1270409
Leverancier: PerkinElmer
Ga naar:
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specificaties
- Voor:SpectrumGX
- Omschrijving:Spacers, PTFE, rectangular, 0,5 mm thickness for semi-demountable cell
- Seq:05