Orders LinkContactUsLinkComponent
Rectangular spacers for SpectrumGX
Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell, SpectrumGX
Catalogus # PERCL1270408
Leverancier:   PerkinElmer
Rectangular spacers for SpectrumGX
Catalogus # PERCL1270408
Leverancier:  PerkinElmer
Rectangular spacers for SpectrumGX
Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell, SpectrumGX
Price
228,00 €
Log in voor beschikbaarheid
1 * 3 ST
Product Details & Documents
PTFE spacers can be used for the analysis of less viscous samples.
Specificaties
  • Voor:
    SpectrumGX
  • Omschrijving:
    Spacers, PTFE, rectangular, 0,2 mm thickness for semi-demountable cell
  • Seq:
    04