Calibration weight sets, class E2, Proof Line
1 g - 1 kg, Without certificate
Catalogus # SARBYCS31-612-AC00
Leverancier: Sartorius Balances
Calibration weight sets, class E2, Proof Line
Catalogus # SARBYCS31-612-AC00
Leverancier: Sartorius Balances
Ga naar:
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Specificaties
- Gewichtset:1 g - 1 kg
- Seq:04
- Type:Without certificate