Calibration weight sets, class E2, Proof Line
1 g - 100 g, Without certificate
Catalog # SARBYCS31-512-AC00
Supplier: Sartorius Balances
Calibration weight sets, class E2, Proof Line
Catalog # SARBYCS31-512-AC00
Supplier: Sartorius Balances
Jump to:
Product Details & Documents
These class E2 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.
Specifications
- Gewichtset:1 g - 100 g
- Seq:02
- Type:Without certificate