Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
QuEChERS dispersive kits
QuEChERS dispersive kits
Buisje met 150 mg primaire/secundaire amine, 45 mg GCB, 900 mg magnesiumsulfaat: verwijdert polaire organische zuren, sommige suikers en lipiden, plus hoge concentraties carotenoïden en chlorofyl, niet voor gebruik met planaire pesticiden, 15 ml
Catalog # HEWL5982-5356
Supplier:   Agilent
QuEChERS dispersive kits
Catalog # HEWL5982-5356
Supplier:  Agilent
QuEChERS dispersive kits
QuEChERS dispersive kits
Buisje met 150 mg primaire/secundaire amine, 45 mg GCB, 900 mg magnesiumsulfaat: verwijdert polaire organische zuren, sommige suikers en lipiden, plus hoge concentraties carotenoïden en chlorofyl, niet voor gebruik met planaire pesticiden, 15 ml
Price
184,00 €
Log in for availability
1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Step 2: Dispersive SPE cleanup
Select the dispersive SPE kit suited to the type of food being analysed and the method you are following. In this step, an aliquot of the sample extract from step one (see QuEChERS extraction kits) is added to a 2 or 15 ml centrifuge tube containing a small amount of SPE sorbent and MgSO₄. The sorbent will pull out interfering matrix materials from the sample, while the MgSO₄ helps remove excess water and improve analyte partitioning.
Specifications
  • Capaciteit:
    15 ml
  • Omschrijving:
    Buisje met 150 mg primaire/secundaire amine, 45 mg GCB, 900 mg magnesiumsulfaat: verwijdert polaire organische zuren, sommige suikers en lipiden, plus hoge concentraties carotenoïden en chlorofyl, niet voor gebruik met planaire pesticiden
  • Seq:
    27
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.