Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
QuEChERS extraction kits

QuEChERS extraction kits

Niet-gebufferd QuEChERS-extractiebuisje, originele QuEChERS-methode, met 4 g magnesiumsulfaat en 1,0 g natriumchloride, voor de analyse van pesticideresiduen in groenten en fruit (monsters van 10 g)
Catalogus # HEWL5982-7550
Leverancier:   Agilent
QuEChERS extraction kits
Catalogus # HEWL5982-7550
Leverancier:  Agilent
QuEChERS extraction kits
QuEChERS extraction kits

Niet-gebufferd QuEChERS-extractiebuisje, originele QuEChERS-methode, met 4 g magnesiumsulfaat en 1,0 g natriumchloride, voor de analyse van pesticideresiduen in groenten en fruit (monsters van 10 g)

Price
543,00 €
Log in voor beschikbaarheid
1 * 200 g
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

Step 1: Extraction
Choose the extraction salt packet based on your method of analysis, AOAC or EN. The buffered extraction salts are amenable for more liable pesticides. Adding solvent and then salts to a comminuted fruit or vegetable sample (10 g or 15 g) enables you to extract the pesticides of interest into the organic layer. However, adding a food sample with a high percentage of water directly to the salts may create an exothermic reaction that can affect your analyte recoveries.

  • Available with or without 50 ml centrifuge tubes and caps
  • Include MgSO₄, NaCl, or other salts for buffering; pre-weighed in anhydrous packet

QuEChERS salts and buffers are packaged in anhydrous packages. This allows you to add them after adding your solvent to the sample, as specified in QuEChERS methodologies.

Specificaties
  • Omschrijving:
    Niet-gebufferd QuEChERS-extractiebuisje, originele QuEChERS-methode, met 4 g magnesiumsulfaat en 1,0 g natriumchloride, voor de analyse van pesticideresiduen in groenten en fruit (monsters van 10 g)
  • Seq:
    15
Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.