Orders LinkContactUsLinkComponent
Drum faucets
Drum faucets
Internal flame arrester, self-closing, wetted PTFE O-rings, 3/4"
Catalogus # 119-0236
Leverancier:   JUSTRITE MANUFACTURING
Drum faucets
Catalogus # 119-0236
Leverancier:  JUSTRITE MANUFACTURING
Drum faucets
Drum faucets
Internal flame arrester, self-closing, wetted PTFE O-rings, 3/4"
Price
73,60 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (PTFE) seal ensures excellent chemical resistance.
  • Safe and easy dispensing
  • FM approved
The faucets feature an internal flame arrester that prevents flashback of fire into the drum for added safety.
Specificaties
  • Schroefdraad:
    3/4"
  • Omschrijving:
    Internal flame arrester, self-closing, wetted PTFE O-rings
  • Seq:
    01