Orders LinkContactUsLinkComponent
Single channel pipettes, variable volume
Single channel pipettes, variable volume
0,2 - 1 ml
Catalog # HACHBBP078
Supplier:   Hach
Single channel pipettes, variable volume
Catalog # HACHBBP078
Supplier:  Hach
Single channel pipettes, variable volume
Single channel pipettes, variable volume
0,2 - 1 ml
Price
537,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These piston pipettes are handy, general-purpose microliter pipettes for transferring exact volumes of liquid. They operate on the principle of air expulsion, making use of disposable tips. The volume that is to be pipetted is shown in a clearly visible display on the pipette handle.
  • Fitted with mechanical tip ejector system to prevent possible contamination
  • Pipette is operated by means of the push button at the end of the handle
  • Factory calibrated; no recalibration is needed for routine use
Specifications
  • Volume:
    0,2 - 1 ml
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Volume
    Voor gebruik met
    Verpakt
    Availability
    Price
  • HACHLYW788
    200 - 1000 µl
    Single channel pipettes BBP078 (0,2 - 1 ml) and BBP163 (1 ml)
    1000 per pack
    Log in for availability
    1 * 1.000 ST - 424,00 €
Product Information
  • Catalog #
    Volume
    Voor gebruik met
    Verpakt
    Availability
    Price
  • HACHBBP079
    200 - 1000 µl
    Single channel pipettes BBP078 and BBP163 (0,2 - 1 ml)
    100 per pack
    Log in for availability
    1 * 100 ST - 53,20 €
Product Information
  • Catalog #
    Volume
    Voor gebruik met
    Verpakt
    Availability
    Price
  • HACHBBP072
    1000 - 5000 µl
    Pipette tips (1 - 5 ml) for BBP065, BBP164 and BBP087
    100 per pack
    Log in for availability
    1 * 100 ST - 80,90 €