Pipette tips, Matrix™ SerialMate™
Standard, 0,5 - 30 µl, +, -, 48,26 mm, Matrix™ SerialMate™, 20 racks, 96 each
Catalogus # MATR5572
Leverancier: THERMO MATRIX TECHNOLOGIES
Pipette tips, Matrix™ SerialMate™
Catalogus # MATR5572
Leverancier: THERMO MATRIX TECHNOLOGIES
Ga naar:
Product Details & Documents
Pipette tips for use with the Matrix™ SerialMate™ pipetting station. Compatible with 96-well plates.
Specificaties
- Verpakt:20 racks, 96 each
- Steriel:+
- Volume:0,5 - 30 µl
- Lengte:48,26 mm
- Tip stijl:Standard
- Voor gebruik met:Matrix™ SerialMate™
- Filter:-
- Cat. No.:MATR5572
- Milieu certificering:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Lage impact op de fabricage