Inserts for storage tubes, 2D Barcoded, Matrix®
Insert for 0,5 ml and 1,0 ml screw top tube caps, paars
Catalogus # 479-1344
Leverancier: THERMO MATRIX TECHNOLOGIES
Inserts for storage tubes, 2D Barcoded, Matrix®
Catalogus # 479-1344
Leverancier: THERMO MATRIX TECHNOLOGIES
Ga naar:
Product Details & Documents
Color inserts snap into cap for a secure fit.
- Inserts are removable
- Free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Specificaties
- Kleur:paars
- Omschrijving:Insert for 0,5 ml and 1,0 ml screw top tube caps
- Seq:01
- Cat. No.:479-1344
- Milieu certificering:NAR Certificate of Retirement
- Environmentally Preferable:Lage impact op de fabricage