Overflow volumetric flasks, class B
432 ml
Catalogus # HECH42115078
Leverancier: GLASWARENFABRIK KARL HECHT
Overflow volumetric flasks, class B
Catalogus # HECH42115078
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
DURAN® borosilicate glass 3.3.
- Acid-proof and alkali-resistant markings
- Very good temperature resistance
- Very good chemical resistance
For water analysis.
Specificaties
- Capaciteit:432 ml
- Seq:12