Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Caffeine, Supelco®

Caffeine, Supelco®

Glass bottle for solids, 5 g
Catalogus # 41019-5G
Irriterend
Leverancier:   Merck
Caffeine, Supelco®
Catalogus # 41019-5G
Leverancier:  Merck
CAS-nummer:  58-08-2
Caffeine, Supelco®
Caffeine, Supelco®

Glass bottle for solids, 5 g

CAS-nummer:  58-08-2
Price
71,10 €
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
Synoniemen:  3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
  • CAS-nummer:58-08-2
  • Formule:C₈H₁₀N₄O₂
  • MDL-nummer:MFCD00005758
  • Molecular Weight:194.19 g/mol
  • Merck index:13,01636
  • Smelten Pt:235…239 °C
  • Dichtheid:1,23 g/cm³ (19 °C)
  •  :M-Clarity™ quality level = MQ100
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-362-1

Documents

Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.