Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Petrischalen

Petrischalen

Contact plate, with 3 vents, 55 mm, 14 mm, Steriel, 25×20/bag
N° de catalogue ANICP055NCS1
Fournisseur:   Anicrin
Petrischalen
N° de catalogue ANICP055NCS1
Fournisseur:  Anicrin
Petrischalen
Petrischalen

Contact plate, with 3 vents, 55 mm, 14 mm, Steriel, 25×20/bag

Prix
402,00 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
Spécifications
  • Ø:
    55 mm
  • Verpakt:
    25×20/bag
  • Steriliteit:
    Steriel
  • Uitvoering:
    Contact plate, with 3 vents
  • Hoogte:
    14 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.