Labels, hazard warning
Hazard labels: danger, confined space, STN 823-297X148-B7541, Polyester, zwart/geel/wit, 148×297 mm, 1 label/card
Catalogus # BRDY223732
Leverancier: Brady
Ga naar:
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certificeringen: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Documents
Specificaties
- Kleur:zwart/geel/wit
- Omschrijving:Hazard labels: danger, confined space
- Afmetingen:148×297 mm
- Materiaal:Polyester
- Seq:32
- Model:STN 823-297X148-B7541
- Verpakt:1 label/card