Staining dishes
Staining set for histology and cytology
Catalogus # BOPT10-10
Leverancier: BIO OPTICA
Ga naar:
Product Details & Documents
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
- Resist temperature between 0 to 170 °C
- Can be used with immunohistochemistry procedures
Specificaties
- Omschrijving:Staining set for histology and cytology
- Seq:04
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- Rack for 25 slides with plastic handle