Orders LinkContactUsLinkComponent
Wash bottles, narrow neck, with thread
Wash bottles, narrow neck, with thread
250 ml, GL 25
Catalogus # HECH49102024
Wash bottles, narrow neck, with thread
Catalogus # HECH49102024
Wash bottles, narrow neck, with thread
Wash bottles, narrow neck, with thread
250 ml, GL 25
Price
3,60 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
LDPE, translucent.
  • Good chemical resistance
  • Good temperature resistance
Specificaties
  • Schroefdraad:
    GL 25
  • Capaciteit:
    250 ml