0,00 €
Bestelling invoeren
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
Orders Link
ContactUsLinkComponent
Flessen
Spuitflessen
Wash bottles, narrow neck, with thread
Wash bottles, narrow neck, with thread
100 ml, GL 18
HECH49102018
:
GLASWARENFABRIK KARL HECHT
Wash bottles, narrow neck, with thread
HECH49102018
:
GLASWARENFABRIK KARL HECHT
Wash bottles, narrow neck, with thread
100 ml, GL 18
2,60 €
1 * 1 ST
-
+
LDPE, translucent.
Good chemical resistance
Good temperature resistance
HECH49102018_TDS_EN
HECH49102024_TDS_EN
HECH49102030_TDS_EN
HECH49102036_TDS_EN
Schroefdraad:
GL 18
Capaciteit:
100 ml
Desiccator lids, DURAN®
#: 467-1485
: DWK Life Sciences
Bufferoplossingen, Certipur® met kleurcode, Supelco®
#: 1.09477.0500
: Merck