Anisool 99%, Reagent Grade
Glass bottle, 1 L
Anisool 99%, Reagent Grade
123226-1L
: 100-66-3
- ≥95% for general laboratory use.
: Methyl phenyl ether, Methoxybenzene
- :100-66-3
- :C₇H₈O
- :MFCD00008354
- :108.14 g/mol
- :13,00672
- :154 °C (1,013 hPa)
- :–37 °C
- :-37 °C
- :1,00 g/cm³ (20 °C)
- :43 °C
- :Kamertemperatuur
- :202-876-1
- :2222
- :3,III
- Vapor Density:3.7 (vs air)
- Assay (GC):Min. 98.5 %
- Identity (IR):complying
- Vapor Pressure:10 mmHg ( 42.2 °C)
- Autoignition Temperature:887 °F
- Verpakking:Glass bottle
- VPE:1 L
- Cat. No.:123226-1L
- Milieu certificering:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Veiligere gezondheid van mens en milieu



