Orders LinkContactUsLinkComponent
dNTP solutions, illustra™
dNTP solutions, illustra™
dCTP, 100 mMol solution, 100 µmol
Catalogus # 28-4065-12
Leverancier:   Cytiva
dNTP solutions, illustra™
Catalogus # 28-4065-12
Leverancier:  Cytiva
CAS-nummer:  
dNTP solutions, illustra™
dNTP solutions, illustra™
dCTP, 100 mMol solution, 100 µmol
CAS-nummer:  
Price
170,00 €
Log in voor beschikbaarheid
1 * 1 EA
Product Details & Documents
illustra™ dNTPs are high purity solutions for enzymatically sensitive applications, such as nucleic acid amplification, sequencing, labelling, mutagenesis, cDNA synthesis and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Specificaties
Specificaties
  • VPE:
    100 µmol
  • Omschrijving:
    dCTP, 100 mMol solution
Specificaties
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.