Inlaat liners
PTV liner, sintered glass, deactivated, 112 µl, 1,5 mm
Catalogus # HEWL5190-1426
Leverancier: Agilent
Ga naar:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specificaties
- Ø inw.:1,5 mm
- Omschrijving:PTV liner, sintered glass, deactivated, 112 µl
- Seq:61