Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Funnels for drums

Funnels for drums

Drum funnel, PE, with sieve, with Ø 580 mm, and filling capacity 5 l
Catalogus # MSGA2730
Leverancier:   asecos
Funnels for drums
Catalogus # MSGA2730
Leverancier:  asecos
Funnels for drums
Funnels for drums

Drum funnel, PE, with sieve, with Ø 580 mm, and filling capacity 5 l

Price
110,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Drum funnel, PE, with sieve, with Ø 580 mm, and filling capacity 5 l
  • Seq:
    06