Funnels for drums
Drum funnel, steel, with Ø×H 650×135 mm , interior Ø×H 620×135 mm, with filling capacity 30 l
Catalogus # MSGA12125
Leverancier: asecos
Ga naar:
Product Details & Documents
Specificaties
- Omschrijving:Drum funnel, steel, with Ø×H 650×135 mm , interior Ø×H 620×135 mm, with filling capacity 30 l
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.