Universal mats
Universalmatta, kraftig, 100 mattor/paket, 400×460 mm
Catalogus # MSGA9951
Leverancier: asecos
Ga naar:
Product Details & Documents
Universal mats for safe absorption of oil, water, cooling liquids and non-aggressive chemicals.
- Capacity up to 16 times of its own weight
- High rib and rub resistance
- Mats are pre-cut: only take sufficient material as required
Specificaties
- Omschrijving:Universalmatta, kraftig
- l×b:400×460 mm
- Verpakt:100 mattor/paket