Absorbent pillows
Absorbent pillow, 20 pillows/pack, 250×250 mm
Catalogus # MSGA9910
Leverancier: asecos
Ga naar:
Product Details & Documents
Special pillows for safe absorption of acids, alkalis, aggressive chemicals and oil.
Specificaties
- Omschrijving:Absorbent pillow
- l×b:250×250 mm
- Verpakt:20 pillows/pack