Universal mats
Universal mat, standard, 200 mats/pack, 400×460 mm
Catalogus # MSGA11326
Leverancier: asecos
Ga naar:
Product Details & Documents
Universal mats for safe absorption of oil, water, cooling liquids and non-aggressive chemicals.
- Capacity up to 16 times of its own weight
- High rib and rub resistance
- Mats are pre-cut: only take sufficient material as required
Specificaties
- Omschrijving:Universal mat, standard
- l×b:400×460 mm
- Verpakt:200 mats/pack